Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCGGGTCCTGTGGCTCCTACAG[C/T]GGGGAGACAGGAGCAATGGTTCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN7L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATXN7L2 - ataxin 7 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYPL2 - synaptophysin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040709.1 | 537 | Silent Mutation | AGC,AGT | S,S 57 | NP_001035799.1 | |
XM_011541283.2 | 537 | Silent Mutation | AGC,AGT | S,S 57 | XP_011539585.1 | |
XM_011541284.2 | 537 | Silent Mutation | AGC,AGT | S,S 57 | XP_011539586.1 | |
XM_011541285.1 | 537 | Silent Mutation | AGC,AGT | S,S 57 | XP_011539587.1 |