Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTTCACTGCAGGTTTTCAAAA[A/G]GCAGATTCCACATCACCCATCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602733 MIM: 604564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH9A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH9A1 - aldehyde dehydrogenase 9 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000696.3 | 1656 | Silent Mutation | GCC,GCT | A,A 517 | NP_000687.3 | |
XM_011509294.2 | 1656 | Silent Mutation | GCC,GCT | A,A 423 | XP_011507596.1 |
MGST3 - microsomal glutathione S-transferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |