Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGGACAGGCATACTCATCGGAC[C/T]AGTCGGAGCAGTGCTGTACATGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LDLRAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDLRAD1 - low density lipoprotein receptor class A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010978.3 | 532 | Nonsense Mutation | TAG,TGG | *,W 196 | NP_001010978.2 | |
NM_001276392.1 | 532 | Nonsense Mutation | TAG,TGG | *,W 157 | NP_001263321.1 | |
NM_001276393.1 | 532 | Nonsense Mutation | TAG,TGG | *,W 153 | NP_001263322.1 | |
NM_001276394.1 | 532 | Nonsense Mutation | TAG,TGG | *,W 107 | NP_001263323.1 | |
NM_001276395.1 | 532 | UTR 3 | NP_001263324.1 | |||
XM_011541446.1 | 532 | Nonsense Mutation | TAG,TGG | *,W 195 | XP_011539748.1 | |
XM_017001264.1 | 532 | Intron | XP_016856753.1 |