Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTGAAGACGGACAGTGACTCG[C/G]ACCTACAGTGAGGAGGGCAGAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 102771 MIM: 139340 | ||||||||||||||||||||
Literature Links: |
AMPD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMPD2 - adenosine monophosphate deaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257360.1 | 790 | Missense Mutation | CAC,GAC | H,D 193 | NP_001244289.1 | |
NM_001257361.1 | 790 | Missense Mutation | CAC,GAC | H,D 75 | NP_001244290.1 | |
NM_001308170.1 | 790 | Missense Mutation | CAC,GAC | H,D 118 | NP_001295099.1 | |
NM_004037.7 | 790 | Missense Mutation | CAC,GAC | H,D 193 | NP_004028.3 | |
NM_139156.3 | 790 | Missense Mutation | CAC,GAC | H,D 112 | NP_631895.1 | |
NM_203404.1 | 790 | Missense Mutation | CAC,GAC | H,D 74 | NP_981949.1 |
GNAT2 - G protein subunit alpha transducin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |