Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGGTAACTAAAGAAGGGCCTG[G/T]GGGGCCTCATCGAATGCAGAATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602758 MIM: 610568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507670 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507670 - uncharacterized LOC100507670 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PI4KB - phosphatidylinositol 4-kinase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF687 - zinc finger protein 687 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304763.1 | 655 | Missense Mutation | GGG,GTG | G,V 118 | NP_001291692.1 | |
NM_001304764.1 | 655 | Missense Mutation | GGG,GTG | G,V 118 | NP_001291693.1 | |
NM_020832.2 | 655 | Missense Mutation | GGG,GTG | G,V 118 | NP_065883.1 | |
XM_005245366.3 | 655 | Missense Mutation | GGG,GTG | G,V 127 | XP_005245423.1 | |
XM_011509811.2 | 655 | Missense Mutation | GGG,GTG | G,V 118 | XP_011508113.1 | |
XM_011509812.2 | 655 | Missense Mutation | GGG,GTG | G,V 118 | XP_011508114.1 | |
XM_011509813.2 | 655 | Missense Mutation | GGG,GTG | G,V 118 | XP_011508115.1 |