Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTGCAGTGGCTGGACCTCGAAG[A/C]CGTCCTCAGGAGAGGGGGCTATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604013 MIM: 601019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT2 - beta-1,4-galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC24 - coiled-coil domain containing 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC6A9 - solute carrier family 6 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024845.2 | 2237 | Missense Mutation | GGC,GTC | G,V 604 | NP_001020016.1 | |
NM_001261380.1 | 2237 | Missense Mutation | GGC,GTC | G,V 608 | NP_001248309.1 | |
NM_006934.3 | 2237 | Missense Mutation | GGC,GTC | G,V 623 | NP_008865.2 | |
NM_201649.3 | 2237 | Missense Mutation | GGC,GTC | G,V 677 | NP_964012.2 | |
XM_011542017.2 | 2237 | Intron | XP_011540319.1 | |||
XM_017002151.1 | 2237 | Missense Mutation | GGC,GTC | G,V 604 | XP_016857640.1 | |
XM_017002152.1 | 2237 | Missense Mutation | GGC,GTC | G,V 577 | XP_016857641.1 | |
XM_017002153.1 | 2237 | Missense Mutation | GGC,GTC | G,V 566 | XP_016857642.1 |