Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGGAGCGTGAGCACTGTCAG[C/T]GCCGCAAAGAAGTACCTAGGGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603673 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD19 - BTB domain containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCH2 - patched 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166292.1 | 3285 | Silent Mutation | GCA,GCG | A,A 1091 | NP_001159764.1 | |
NM_003738.4 | 3285 | Silent Mutation | GCA,GCG | A,A 1091 | NP_003729.3 |