Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTATGTTCCATTTAGGTGAAGAA[G/A]GCCCTGTGTGTCCTCGTCCAACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR3C - RNA polymerase III subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303456.1 | 279 | Missense Mutation | NP_001290385.1 | |||
NM_006468.7 | 279 | Missense Mutation | NP_006459.3 | |||
XM_005272927.3 | 279 | UTR 5 | XP_005272984.1 | |||
XM_005272928.3 | 279 | UTR 5 | XP_005272985.1 | |||
XM_011509078.2 | 279 | Missense Mutation | XP_011507380.1 |
RNF115 - ring finger protein 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |