Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGCGCTGTATCGTGTCGCCGG[C/T]AGGGAGGCATAGCGCCTCTCTGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPLAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYPLAL1 - lysophospholipase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300769.1 | 51 | Missense Mutation | GCA,GTA | A,V 17 | NP_001287698.1 | |
NM_001300770.1 | 51 | Missense Mutation | GCA,GTA | A,V 17 | NP_001287699.1 | |
NM_001300771.1 | 51 | Intron | NP_001287700.1 | |||
NM_001300772.1 | 51 | Intron | NP_001287701.1 | |||
NM_138794.4 | 51 | Missense Mutation | GCA,GTA | A,V 17 | NP_620149.2 | |
XM_006711162.3 | 51 | Missense Mutation | GCA,GTA | A,V 17 | XP_006711225.1 | |
XM_011509182.2 | 51 | Missense Mutation | GCA,GTA | A,V 17 | XP_011507484.1 | |
XM_011509183.2 | 51 | UTR 5 | XP_011507485.1 | |||
XM_017000271.1 | 51 | Missense Mutation | GCA,GTA | A,V 17 | XP_016855760.1 | |
XM_017000272.1 | 51 | UTR 5 | XP_016855761.1 |
LYPLAL1-AS1 - LYPLAL1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |