Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGGGGCGGGTGCTTTATTGGG[G/T]GCTGGGACTGGGTCCAGGACAGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615679 MIM: 609151 | ||||||||||||||||||||
Literature Links: |
CEP85 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP85 - centrosomal protein 85 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3BGRL3 - SH3 domain binding glutamate rich protein like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031286.3 | 1343 | Intron | NP_112576.1 |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077262.1 | 1343 | Missense Mutation | ACC,CCC | T,P 399 | NP_001070730.1 | |
NM_145345.2 | 1343 | Missense Mutation | ACC,CCC | T,P 486 | NP_663320.2 | |
NM_183008.2 | 1343 | Missense Mutation | ACC,CCC | T,P 519 | NP_892120.2 |