Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCACAATCTTCCCTCGATTTAC[A/G]TGGGAGTTGCATTCCTGGAAAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615406 MIM: 611514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG12-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNG12-AS1 - GNG12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WLS - wntless Wnt ligand secretion mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002292.3 | 1766 | Missense Mutation | CGT,TGT | R,C 509 | NP_001002292.3 | |
NM_001193334.1 | 1766 | Intron | NP_001180263.1 | |||
NM_024911.6 | 1766 | Intron | NP_079187.3 | |||
XM_011542191.2 | 1766 | Missense Mutation | CGT,TGT | R,C 511 | XP_011540493.1 | |
XM_011542192.2 | 1766 | Missense Mutation | CGT,TGT | R,C 466 | XP_011540494.1 | |
XM_017002390.1 | 1766 | Intron | XP_016857879.1 |