Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTCTGAGTTGGGGGAAGGCCCC[A/G]GAGACCTCCAGCCCATCCATCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611748 | ||||||||||||||||||||
Literature Links: |
MTMR11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTMR11 - myotubularin related protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTUD7B - OTU deubiquitinase 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020205.3 | 2475 | Missense Mutation | CGG,TGG | R,W 793 | NP_064590.2 | |
XM_011509782.1 | 2475 | Missense Mutation | CGG,TGG | R,W 802 | XP_011508084.1 | |
XM_011509783.2 | 2475 | Missense Mutation | CGG,TGG | R,W 802 | XP_011508085.1 | |
XM_011509784.2 | 2475 | Missense Mutation | CGG,TGG | R,W 802 | XP_011508086.1 | |
XM_011509785.2 | 2475 | Missense Mutation | CGG,TGG | R,W 802 | XP_011508087.1 | |
XM_011509786.1 | 2475 | Missense Mutation | CGG,TGG | R,W 723 | XP_011508088.1 | |
XM_011509787.1 | 2475 | Missense Mutation | CGG,TGG | R,W 723 | XP_011508089.1 | |
XM_011509788.1 | 2475 | Missense Mutation | CGG,TGG | R,W 723 | XP_011508090.1 | |
XM_017001850.1 | 2475 | Missense Mutation | CGG,TGG | R,W 714 | XP_016857339.1 | |
XM_017001851.1 | 2475 | Missense Mutation | CGG,TGG | R,W 664 | XP_016857340.1 |