Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATCTGTGGCGCGATTATCTGTAC[C/G]CAAAACAGTATGAGTGGGTCCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCRTR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCRTR1 - hypocretin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001525.2 | 1000 | Missense Mutation | CCA,GCA | P,A 42 | NP_001516.2 | |
XM_017001105.1 | 1000 | Missense Mutation | CCA,GCA | P,A 42 | XP_016856594.1 | |
XM_017001106.1 | 1000 | Missense Mutation | CCA,GCA | P,A 42 | XP_016856595.1 | |
XM_017001107.1 | 1000 | Missense Mutation | CCA,GCA | P,A 42 | XP_016856596.1 |