Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGTCTGCAGCTTCACGGGGAGG[C/T]TGGGTGCTAAACAGCACACGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NGF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NGF - nerve growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002506.2 | 445 | Silent Mutation | CAA,CAG | Q,Q 92 | NP_002497.2 | |
XM_006710663.3 | 445 | Silent Mutation | CAA,CAG | Q,Q 92 | XP_006710726.1 | |
XM_011541518.2 | 445 | Silent Mutation | CAA,CAG | Q,Q 147 | XP_011539820.1 |