Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCGACAGCCACAAGCTTGTCTT[C/G]AACGTGCCCTTTTCTGATTCCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PADI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PADI1 - peptidyl arginine deiminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013358.2 | 598 | Missense Mutation | TTC,TTG | F,L 201 | NP_037490.2 | |
XM_011541307.1 | 598 | Missense Mutation | TTC,TTG | F,L 201 | XP_011539609.1 | |
XM_017001101.1 | 598 | Missense Mutation | TTC,TTG | F,L 150 | XP_016856590.1 | |
XM_017001102.1 | 598 | Missense Mutation | TTC,TTG | F,L 150 | XP_016856591.1 | |
XM_017001103.1 | 598 | Missense Mutation | TTC,TTG | F,L 201 | XP_016856592.1 |