Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGTTCTTACCAGTAGGGTGTA[G/A]GCAGTTGGGCTGAGGGTATAGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 116890 | ||||||||||||||||||||
Literature Links: |
CTSE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTSE - cathepsin E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317331.1 | 995 | Missense Mutation | CCT,CTT | P,L 216 | NP_001304260.1 | |
NM_001910.3 | 995 | Silent Mutation | GCC,GCT | A,A 338 | NP_001901.1 | |
NM_148964.2 | 995 | Missense Mutation | CCT,CTT | P,L 291 | NP_683865.1 | |
XM_011509244.1 | 995 | Silent Mutation | GCC,GCT | A,A 343 | XP_011507546.1 | |
XM_011509245.1 | 995 | Missense Mutation | CCT,CTT | P,L 296 | XP_011507547.1 |
RAB7B - RAB7B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |