Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGAAAAACAAGGCCTGTTCAAA[A/C]ACAAGACAACTTCCTCTCACTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111680 MIM: 605348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHD - Rh blood group D antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127691.2 | 1348 | UTR 3 | NP_001121163.1 | |||
NM_001282867.1 | 1348 | UTR 3 | NP_001269796.1 | |||
NM_001282868.1 | 1348 | Missense Mutation | AAA,ACA | K,T 361 | NP_001269797.1 | |
NM_001282869.1 | 1348 | Missense Mutation | AAA,ACA | K,T 379 | NP_001269798.1 | |
NM_001282870.1 | 1348 | Missense Mutation | AAC,CAC | N,H 406 | NP_001269799.1 | |
NM_001282871.1 | 1348 | Missense Mutation | AAC,CAC | N,H 436 | NP_001269800.1 | |
NM_001282872.1 | 1348 | UTR 3 | NP_001269801.1 | |||
NM_016124.4 | 1348 | UTR 3 | NP_057208.2 | |||
XM_017002015.1 | 1348 | Intron | XP_016857504.1 |
RSRP1 - arginine and serine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321772.1 | 1348 | Intron | NP_001308701.1 | |||
NM_020317.4 | 1348 | Intron | NP_064713.3 | |||
XM_011541797.1 | 1348 | Intron | XP_011540099.1 |
TMEM50A - transmembrane protein 50A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |