Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGGGAAGGTGAGGCGCGGTGA[C/G]ATGTACAGCCAGCCATTGTCAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf24 - chromosome 20 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLA2 - Src like adaptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032214.3 | 894 | Missense Mutation | ATC,ATG | I,M 160 | NP_115590.1 | |
NM_175077.2 | 894 | Missense Mutation | ATC,ATG | I,M 160 | NP_778252.1 | |
XM_017028098.1 | 894 | Missense Mutation | ATC,ATG | I,M 160 | XP_016883587.1 |
TGIF2-C20orf24 - TGIF2-C20orf24 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |