Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTGCGGTGGCCTGGAGAAGT[C/T]GGGGGCCTCCCCATGACAGAGCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605704 | ||||||||||||||||||||
Literature Links: |
APCDD1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APCDD1L - APC down-regulated 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304787.1 | 1612 | Missense Mutation | AAC,GAC | N,D 472 | NP_001291716.1 | |
NM_153360.2 | 1612 | Missense Mutation | AAC,GAC | N,D 461 | NP_699191.1 |
VAPB - VAMP associated protein B and C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |