Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAGCTGCAGGCTCCTCAGTCTC[A/G]TGTGATTCTAAAGTGCTTGCATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRMT6 - tRNA methyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281467.1 | 1689 | Silent Mutation | CAC,CAT | H,H 310 | NP_001268396.1 | |
NM_015939.4 | 1689 | Silent Mutation | CAC,CAT | H,H 480 | NP_057023.2 | |
XM_017027875.1 | 1689 | Silent Mutation | CAC,CAT | H,H 298 | XP_016883364.1 |