Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGTGGTGGGGGAGCAGGGTCA[C/T]TGCAGCTCCTTGAGCAGCTGGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303457.1 | 3373 | Silent Mutation | NP_001290386.1 | |||
NM_014657.2 | 3373 | Silent Mutation | NP_055472.1 | |||
XM_011529114.1 | 3373 | Intron | XP_011527416.1 | |||
XM_017028148.1 | 3373 | Intron | XP_016883637.1 |