Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGCCACAGGCTCCTCTGGGGTC[C/T]GCTCCTTGGGCTTTCGGCCCCGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UCKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UCKL1-AS1 - UCKL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF512B - zinc finger protein 512B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020713.2 | 2683 | Missense Mutation | CAG,CGG | Q,R 851 | NP_065764.1 | |
XM_005260226.3 | 2683 | Missense Mutation | CAG,CGG | Q,R 865 | XP_005260283.1 | |
XM_011528929.2 | 2683 | Missense Mutation | CAG,CGG | Q,R 851 | XP_011527231.1 | |
XM_011528930.2 | 2683 | Missense Mutation | CAG,CGG | Q,R 832 | XP_011527232.1 | |
XM_011528931.2 | 2683 | Missense Mutation | CAG,CGG | Q,R 521 | XP_011527233.1 | |
XM_011528932.2 | 2683 | Missense Mutation | CAG,CGG | Q,R 520 | XP_011527234.1 | |
XM_011528933.2 | 2683 | Intron | XP_011527235.1 |