Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTGTAGCTGGTTCGCTCTACTG[C/T]AAGGGGCAGGACGGTTTTACAGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C21orf62 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C21orf62 - chromosome 21 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162495.2 | 505 | Missense Mutation | ACA,GCA | T,A 70 | NP_001155967.2 | |
NM_001162496.2 | 505 | Missense Mutation | ACA,GCA | T,A 70 | NP_001155968.2 | |
NM_019596.5 | 505 | Missense Mutation | ACA,GCA | T,A 70 | NP_062542.5 | |
XM_011529653.2 | 505 | Intron | XP_011527955.1 |
C21orf62-AS1 - C21orf62 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |