Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCTGGTTATTCTTCCTGAAAT[A/G]AGTTTGGTGATTAGAAATCTGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM3AP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003906.4 | 103 | Intron | NP_003897.2 | |||
XM_005261203.4 | 103 | Intron | XP_005261260.1 | |||
XM_005261204.4 | 103 | Intron | XP_005261261.1 | |||
XM_005261205.3 | 103 | Intron | XP_005261262.1 |
YBEY - ybeY metallopeptidase (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006114.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001006114.1 | |
NM_001314022.1 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001300951.1 | |
NM_001314023.1 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001300952.1 | |
NM_001314024.1 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001300953.1 | |
NM_001314025.1 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001300954.1 | |
NM_001314026.1 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_001300955.1 | |
NM_058181.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | NP_478061.1 | |
XM_005261156.3 | 103 | Missense Mutation | ATA,ATG | I,M 130 | XP_005261213.2 | |
XM_011529629.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | XP_011527931.1 | |
XM_011529630.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | XP_011527932.1 | |
XM_011529631.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | XP_011527933.1 | |
XM_011529633.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | XP_011527935.1 | |
XM_011529634.2 | 103 | Missense Mutation | ATA,ATG | I,M 130 | XP_011527936.2 | |
XM_011529635.2 | 103 | Missense Mutation | ATA,ATG | I,M 1 | XP_011527937.1 | |
XM_017028394.1 | 103 | Missense Mutation | ATA,ATG | I,M 130 | XP_016883883.1 | |
XM_017028395.1 | 103 | Intron | XP_016883884.1 | |||
XM_017028396.1 | 103 | Intron | XP_016883885.1 |