Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGGGCCTCCACACGGCCTCCTC[C/T]AGGCGCGCGGAGGGCGCCATGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01426 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01426 - long intergenic non-protein coding RNA 1426 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUNX1 - runt related transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001890.2 | 1411 | Silent Mutation | CTA,CTG | L,L 445 | NP_001001890.1 | |
NM_001122607.1 | 1411 | Intron | NP_001116079.1 | |||
NM_001754.4 | 1411 | Silent Mutation | CTA,CTG | L,L 472 | NP_001745.2 | |
XM_005261068.3 | 1411 | Silent Mutation | CTA,CTG | L,L 460 | XP_005261125.1 | |
XM_005261069.4 | 1411 | Silent Mutation | CTA,CTG | L,L 408 | XP_005261126.1 | |
XM_011529766.2 | 1411 | Silent Mutation | CTA,CTG | L,L 472 | XP_011528068.1 | |
XM_011529767.2 | 1411 | Silent Mutation | CTA,CTG | L,L 459 | XP_011528069.1 | |
XM_011529768.2 | 1411 | Silent Mutation | CTA,CTG | L,L 395 | XP_011528070.1 | |
XM_011529770.2 | 1411 | Intron | XP_011528072.1 | |||
XM_017028487.1 | 1411 | Silent Mutation | CTA,CTG | L,L 421 | XP_016883976.1 |