Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCGGCAGGGGGGTTGGAGGAGG[A/C]GGCAGAGTTCACCCTGCGAGGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611520 | ||||||||||||||||||||
Literature Links: |
POLDIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLDIP3 - DNA polymerase delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278657.1 | 1258 | Missense Mutation | GCC,TCC | A,S 402 | NP_001265586.1 | |
NM_032311.4 | 1258 | Missense Mutation | GCC,TCC | A,S 385 | NP_115687.2 | |
NM_178136.2 | 1258 | Missense Mutation | GCC,TCC | A,S 356 | NP_835237.1 | |
XM_011530457.1 | 1258 | UTR 3 | XP_011528759.1 | |||
XM_017029010.1 | 1258 | UTR 3 | XP_016884499.1 | |||
XM_017029011.1 | 1258 | Intron | XP_016884500.1 |
RRP7BP - ribosomal RNA processing 7 homolog B, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |