Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGAGCAGGGCGGGGGCCTCCA[C/G]GGGAGACAGCTGAGAGCGAGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611230 | ||||||||||||||||||||
Literature Links: |
LMF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMF2 - lipase maturation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033200.2 | 2374 | Missense Mutation | CTG,GTG | L,V 633 | NP_149977.2 | |
XM_005261955.3 | 2374 | Missense Mutation | CTG,GTG | L,V 608 | XP_005262012.1 | |
XM_006724426.3 | 2374 | Missense Mutation | CTG,GTG | L,V 578 | XP_006724489.1 | |
XM_006724427.3 | 2374 | Missense Mutation | CTG,GTG | L,V 578 | XP_006724490.1 | |
XM_017029077.1 | 2374 | Missense Mutation | CTG,GTG | L,V 595 | XP_016884566.1 |
NCAPH2 - non-SMC condensin II complex subunit H2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |