Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCTCAAGGGTGCAACAAAAC[A/G]TTCCAAGTGGGACAGATACTGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOL1 - apolipoprotein L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136540.1 | Intron | NP_001130012.1 | ||||
NM_001136541.1 | Intron | NP_001130013.1 | ||||
NM_003661.3 | Intron | NP_003652.2 | ||||
NM_145343.2 | Intron | NP_663318.1 | ||||
XM_005261796.3 | Intron | XP_005261853.1 | ||||
XM_011530478.2 | Intron | XP_011528780.1 |