Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGCTCGTGCCCGCAATTTGTT[C/T]ACTAGTTCTACCACACTGCAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607252 | ||||||||||||||||||||
Literature Links: |
APOL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOL2 - apolipoprotein L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030882.3 | 993 | Intron | NP_112092.2 | |||
NM_145637.2 | 993 | Silent Mutation | GTA,GTG | V,V 172 | NP_663612.2 | |
XM_011530074.1 | 993 | Intron | XP_011528376.1 | |||
XM_011530075.1 | 993 | Intron | XP_011528377.1 | |||
XM_011530076.2 | 993 | Intron | XP_011528378.1 | |||
XM_011530077.2 | 993 | Intron | XP_011528379.1 | |||
XM_011530078.1 | 993 | Intron | XP_011528380.1 | |||
XM_017028724.1 | 993 | Intron | XP_016884213.1 |