Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATATTCTTTGTGTGTGACACCC[A/G]TAGGCCAGTCAATGTCGTCAATGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603465 MIM: 601754 | ||||||||||||||||||||
Literature Links: |
CDC45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC45 - cell division cycle 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178010.2 | 445 | Missense Mutation | CAT,CGT | H,R 101 | NP_001171481.1 | |
NM_001178011.2 | 445 | Intron | NP_001171482.1 | |||
NM_003504.4 | 445 | Missense Mutation | CAT,CGT | H,R 101 | NP_003495.1 | |
XM_005261285.2 | 445 | Missense Mutation | CAT,CGT | H,R 101 | XP_005261342.1 | |
XM_005261286.3 | 445 | Missense Mutation | CAT,CGT | H,R 101 | XP_005261343.1 | |
XM_011530415.1 | 445 | Missense Mutation | CAT,CGT | H,R 101 | XP_011528717.1 | |
XM_011530416.1 | 445 | Missense Mutation | CAT,CGT | H,R 89 | XP_011528718.1 | |
XM_011530417.2 | 445 | Intron | XP_011528719.1 | |||
XM_011530418.2 | 445 | Intron | XP_011528720.1 | |||
XM_017028966.1 | 445 | Intron | XP_016884455.1 | |||
XM_017028967.1 | 445 | Intron | XP_016884456.1 |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |