Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCTTCAGCTTCTCAGGTGGAA[C/T]GTAATCTCCTTCCTTCTCGGTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602300 MIM: 605819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAL3ST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAL3ST1 - galactose-3-O-sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PES1 - pescadillo ribosomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243225.1 | 1855 | Intron | NP_001230154.1 | |||
NM_001282327.1 | 1855 | Missense Mutation | ATT,GTT | I,V 296 | NP_001269256.1 | |
NM_001282328.1 | 1855 | Missense Mutation | ATT,GTT | I,V 296 | NP_001269257.1 | |
NM_014303.3 | 1855 | Intron | NP_055118.1 | |||
XM_006724173.1 | 1855 | Missense Mutation | ATT,GTT | I,V 435 | XP_006724236.1 | |
XM_017028678.1 | 1855 | Missense Mutation | ATT,GTT | I,V 296 | XP_016884167.1 |