Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCACAGTGGAATCATCAAACGG[A/T]CTTCCTTCTCATAGATGTCAGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP18 - dual specificity phosphatase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304794.1 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | NP_001291723.1 | |
NM_001304795.1 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | NP_001291724.1 | |
NM_001304796.1 | 1287 | UTR 3 | NP_001291725.1 | |||
NM_152511.4 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | NP_689724.3 | |
XM_005261368.4 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_005261425.1 | |
XM_006724148.3 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_006724211.1 | |
XM_011529920.2 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_011528222.1 | |
XM_011529921.2 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_011528223.1 | |
XM_017028627.1 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_016884116.1 | |
XM_017028628.1 | 1287 | Missense Mutation | GAC,GTC | D,V 182 | XP_016884117.1 |
SLC35E4 - solute carrier family 35 member E4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001479.3 | 1287 | Intron | NP_001001479.1 | |||
NM_001318370.1 | 1287 | Intron | NP_001305299.1 | |||
NM_001318371.1 | 1287 | Intron | NP_001305300.1 | |||
XM_017028794.1 | 1287 | Intron | XP_016884283.1 | |||
XM_017028795.1 | 1287 | Intron | XP_016884284.1 |