Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACCCTTGCACTCTGTGTCCACA[C/T]GGTGGTCGTAGTAGCGCAGCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610877 MIM: 603560 | ||||||||||||||||||||
Literature Links: |
PPP6R2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP6R2 - protein phosphatase 6 regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBF1 - SET binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002972.3 | 5702 | Missense Mutation | CAT,CGT | H,R 1824 | NP_002963.2 | |
XM_005261935.2 | 5702 | Missense Mutation | CAT,CGT | H,R 1798 | XP_005261992.1 | |
XM_011530709.2 | 5702 | Intron | XP_011529011.1 | |||
XM_011530710.2 | 5702 | Intron | XP_011529012.1 | |||
XM_017028905.1 | 5702 | Intron | XP_016884394.1 |