Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTACACAAAATATATTTATGTCA[A/C]GTAAGTATTTTTTCTATTTGAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GMCL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMCL1 - germ cell-less, spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178439.4 | 617 | Intron | NP_848526.1 | |||
XM_011533033.1 | 617 | Intron | XP_011531335.1 | |||
XM_011533034.2 | 617 | UTR 5 | XP_011531336.1 | |||
XM_017004704.1 | 617 | Intron | XP_016860193.1 | |||
XM_017004705.1 | 617 | Intron | XP_016860194.1 |