Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACGGTGGCACACATTGCACACA[A/T]AGGGTTTGGCCTCACTGTGCTTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605939 MIM: 604083 | ||||||||||||||||||||
Literature Links: |
PLCD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCD4 - phospholipase C delta 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF142 - zinc finger protein 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105537.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1593 | NP_001099007.1 | |
XM_006712730.3 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_006712793.1 | |
XM_011511784.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_011510086.1 | |
XM_011511785.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_011510087.1 | |
XM_011511786.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_011510088.1 | |
XM_011511787.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_011510089.1 | |
XM_011511788.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_011510090.1 | |
XM_011511789.2 | 5986 | Missense Mutation | TAT,TTT | Y,F 1593 | XP_011510091.1 | |
XM_017004871.1 | 5986 | Missense Mutation | TAT,TTT | Y,F 1793 | XP_016860360.1 | |
XM_017004872.1 | 5986 | Missense Mutation | TAT,TTT | Y,F 1593 | XP_016860361.1 |