Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTGTTGTTTACAGAAACAAAC[C/T]GGAAGCAGTAGAAGTAACATTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPC2 - actin related protein 2/3 complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005731.3 | 157 | Missense Mutation | CCG,CTG | P,L 28 | NP_005722.1 | |
NM_152862.2 | 157 | Missense Mutation | CCG,CTG | P,L 28 | NP_690601.1 | |
XM_011510471.1 | 157 | Intron | XP_011508773.1 | |||
XM_017003113.1 | 157 | UTR 5 | XP_016858602.1 | |||
XM_017003114.1 | 157 | UTR 5 | XP_016858603.1 | |||
XM_017003115.1 | 157 | UTR 5 | XP_016858604.1 |