Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTGTTATGCAGCCTAGTTAAGC[G/T]TCTCACGCTGAATCTGGTAGTAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606352 MIM: 606575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALS2 - ALS2, alsin Rho guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135745.1 | 4612 | Intron | NP_001129217.1 | |||
NM_020919.3 | 4612 | Missense Mutation | AAG,ACG | K,T 1655 | NP_065970.2 | |
XM_006712654.2 | 4612 | Missense Mutation | AAG,ACG | K,T 1655 | XP_006712717.1 | |
XM_006712655.3 | 4612 | Missense Mutation | AAG,ACG | K,T 967 | XP_006712718.1 | |
XM_011511530.2 | 4612 | Missense Mutation | AAG,ACG | K,T 1542 | XP_011509832.1 | |
XM_017004569.1 | 4612 | Missense Mutation | AAG,ACG | K,T 1654 | XP_016860058.1 | |
XM_017004570.1 | 4612 | Intron | XP_016860059.1 | |||
XM_017004571.1 | 4612 | Missense Mutation | AAG,ACG | K,T 966 | XP_016860060.1 | |
XM_017004572.1 | 4612 | Missense Mutation | AAG,ACG | K,T 861 | XP_016860061.1 |
MPP4 - membrane palmitoylated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |