Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAAAATGCCAGTTTGACCTCG[A/G]AGTTTTCCTTCCCACCAATCAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SH3YL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH3YL1 - SH3 and SYLF domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159597.2 | 1037 | Silent Mutation | CTC,CTT | L,L 308 | NP_001153069.1 | |
NM_001282682.1 | 1037 | Silent Mutation | CTC,CTT | L,L 212 | NP_001269611.1 | |
NM_001282687.1 | 1037 | Silent Mutation | CTC,CTT | L,L 231 | NP_001269616.1 | |
NM_015677.3 | 1037 | Silent Mutation | CTC,CTT | L,L 327 | NP_056492.2 |