Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCCAATCGTGTACTCGCCGCG[A/C]TACAACATCACCTTCATGGGCCTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607226 | |||||||||||||||||||||||
Literature Links: |
HDAC11 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HDAC11 - histone deacetylase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136041.2 | 290 | UTR 5 | NP_001129513.1 | |||
NM_024827.3 | 290 | Silent Mutation | CGA,CGC | R,R 23 | NP_079103.2 | |
XM_005265491.1 | 290 | UTR 5 | XP_005265548.1 | |||
XM_005265492.1 | 290 | Silent Mutation | CGA,CGC | R,R 23 | XP_005265549.1 | |
XM_006713338.1 | 290 | Silent Mutation | CGA,CGC | R,R 23 | XP_006713401.1 | |
XM_011534131.1 | 290 | UTR 5 | XP_011532433.1 | |||
XM_011534132.2 | 290 | UTR 5 | XP_011532434.2 | |||
XM_011534135.1 | 290 | UTR 5 | XP_011532437.1 | |||
XM_017007234.1 | 290 | UTR 5 | XP_016862723.1 |
HDAC11-AS1 - HDAC11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |