Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGCGGTCTCTGCACTAGCTCACA[C/T]TGCTCCGGGCCGTCCGCTGGGCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607082 MIM: 607068 MIM: 607069 | ||||||||||||||||||||
Literature Links: |
CACNA2D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA2D2 - calcium voltage-gated channel auxiliary subunit alpha2delta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005505.2 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1077 | NP_001005505.1 | |
NM_001174051.2 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1082 | NP_001167522.1 | |
NM_001291101.1 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1008 | NP_001278030.1 | |
NM_006030.3 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1075 | NP_006021.2 | |
XM_005265581.4 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1076 | XP_005265638.1 | |
XM_011534243.2 | 3201 | Silent Mutation | CAA,CAG | Q,Q 1083 | XP_011532545.1 |
CYB561D2 - cytochrome b561 family member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291284.1 | 3201 | Intron | NP_001278213.1 | |||
NM_007022.4 | 3201 | Intron | NP_008953.1 |
TMEM115 - transmembrane protein 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |