Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACAGCGTGGTCACTGGACGAG[A/G]CCGAATCAATGGAAGATTGGTTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 232050 | ||||||||||||||||||||
Literature Links: |
PCCB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCCB - propionyl-CoA carboxylase beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000532.4 | 386 | Missense Mutation | GAC,GGC | D,G 112 | NP_000523.2 | |
NM_001178014.1 | 386 | Missense Mutation | GAC,GGC | D,G 112 | NP_001171485.1 | |
XM_011512873.1 | 386 | Missense Mutation | GAC,GGC | D,G 112 | XP_011511175.1 |