Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCATCAAGCTTCCCTTCTTCA[A/T]CATCCTCTCCTTAATGGCTGGGGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615601 | |||||||||||||||||||||||
Literature Links: |
GADL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GADL1 - glutamate decarboxylase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207359.2 | 1467 | Missense Mutation | ATG,TTG | M,L 474 | NP_997242.2 | |
XM_017006297.1 | 1467 | Missense Mutation | ATG,TTG | M,L 455 | XP_016861786.1 | |
XM_017006298.1 | 1467 | Missense Mutation | ATG,TTG | M,L 228 | XP_016861787.1 |