Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCACGGTATTGCCCCAGATGTC[A/G]TACTGAGGCTTTCGTGCCCCTATCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600293 | |||||||||||||||||||||||
Literature Links: |
ADCY5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ADCY5 - adenylate cyclase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199642.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 846 | NP_001186571.1 | |
NM_183357.2 | 2684 | Silent Mutation | TAC,TAT | Y,Y 1196 | NP_899200.1 | |
XM_005247077.3 | 2684 | Silent Mutation | TAC,TAT | Y,Y 1221 | XP_005247134.1 | |
XM_005247078.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 871 | XP_005247135.1 | |
XM_006713483.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 854 | XP_006713546.1 | |
XM_006713484.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 780 | XP_006713547.1 | |
XM_011512359.2 | 2684 | Silent Mutation | TAC,TAT | Y,Y 888 | XP_011510661.1 | |
XM_011512360.2 | 2684 | Silent Mutation | TAC,TAT | Y,Y 858 | XP_011510662.1 | |
XM_011512361.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 780 | XP_011510663.1 | |
XM_017005638.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 855 | XP_016861127.1 | |
XM_017005639.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 855 | XP_016861128.1 | |
XM_017005640.1 | 2684 | Silent Mutation | TAC,TAT | Y,Y 780 | XP_016861129.1 |