Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGCCACACACTGGCATTGCT[A/G]TTGTCCAAGAGAGAAATTCCCGACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606867 MIM: 611430 MIM: 612167 | |||||||||||||||||||||||
Literature Links: |
GORASP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GORASP1 - golgi reassembly stacking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278789.1 | 1696 | Silent Mutation | AAC,AAT | N,N 223 | NP_001265718.1 | |
NM_001278790.1 | 1696 | Silent Mutation | AAC,AAT | N,N 163 | NP_001265719.1 | |
NM_031899.3 | 1696 | Silent Mutation | AAC,AAT | N,N 318 | NP_114105.1 | |
XM_006713301.3 | 1696 | Silent Mutation | AAC,AAT | N,N 323 | XP_006713364.1 | |
XM_011534020.2 | 1696 | Silent Mutation | AAC,AAT | N,N 293 | XP_011532322.1 | |
XM_011534021.2 | 1696 | Silent Mutation | AAC,AAT | N,N 263 | XP_011532323.1 | |
XM_017007050.1 | 1696 | Silent Mutation | AAC,AAT | N,N 288 | XP_016862539.1 | |
XM_017007051.1 | 1696 | Silent Mutation | AAC,AAT | N,N 258 | XP_016862540.1 | |
XM_017007052.1 | 1696 | Silent Mutation | AAC,AAT | N,N 161 | XP_016862541.1 |
TTC21A - tetratricopeptide repeat domain 21A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR48 - WD repeat domain 48 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |