Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTCAGTGAACCACCTGACCAC[A/G]AGAAGAAAACCAACCAAGAAGTGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614788 | |||||||||||||||||||||||
Literature Links: |
FGD5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FGD5 - FYVE, RhoGEF and PH domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320276.1 | 1008 | Missense Mutation | AAG,GAG | K,E 300 | NP_001307205.1 | |
NM_152536.3 | 1008 | Missense Mutation | AAG,GAG | K,E 300 | NP_689749.3 | |
XM_011533422.1 | 1008 | Missense Mutation | AAG,GAG | K,E 300 | XP_011531724.1 | |
XM_011533423.1 | 1008 | Missense Mutation | AAG,GAG | K,E 300 | XP_011531725.1 |
LOC152274 - uncharacterized LOC152274 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |