Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCCTCCAGCCTTCTTCACCA[C/T]GTCCAAGATGCCGGCCAAGAAGAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TSC22D2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TSC22D2 - TSC22 domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303264.1 | 1018 | Missense Mutation | ACG,ATG | T,M 1 | NP_001290193.1 | |
NM_014779.3 | 1018 | Missense Mutation | ACG,ATG | T,M 1 | NP_055594.1 | |
XM_011513337.2 | 1018 | Missense Mutation | ACG,ATG | T,M 1 | XP_011511639.1 |