Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTGAATATCTGCCATACCTTTT[C/T]CCCAAACTCTCTTGCTTTAATTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604371 MIM: 609929 | ||||||||||||||||||||
Literature Links: |
HHLA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HHLA2 - HERV-H LTR-associating 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH15 - myosin heavy chain 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014981.1 | 5999 | Missense Mutation | AAA,GAA | K,E 1941 | NP_055796.1 | |
XM_011512559.2 | 5999 | Missense Mutation | AAA,GAA | K,E 1941 | XP_011510861.1 | |
XM_017005922.1 | 5999 | Missense Mutation | AAA,GAA | K,E 1594 | XP_016861411.1 |