Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGTAATCCCAAGAATGGTGAT[C/G]TCGCCAATATTGAAGGCATCTTGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602304 | |||||||||||||||||||||||
Literature Links: |
NR1D2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NR1D2 - nuclear receptor subfamily 1 group D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145425.1 | 497 | UTR 5 | NP_001138897.1 | |||
NM_005126.4 | 497 | Missense Mutation | CTC,GTC | L,V 60 | NP_005117.3 | |
XM_006713451.3 | 497 | Missense Mutation | CTC,GTC | L,V 60 | XP_006713514.1 |