Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGGGAATTTCTTCAGAAAGACA[A/G]TATCAGCGTGCATTATTTCTGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603089 | ||||||||||||||||||||
Literature Links: |
BAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAP1 - BRCA1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF7 - PHD finger protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278221.2 | 920 | Missense Mutation | AAT,AGT | N,S 54 | NP_001265150.1 | |
NM_001321126.1 | 920 | Missense Mutation | AAT,AGT | N,S 54 | NP_001308055.1 | |
NM_001321127.1 | 920 | Missense Mutation | AAT,AGT | N,S 54 | NP_001308056.1 | |
NM_016483.6 | 920 | Missense Mutation | AAT,AGT | N,S 54 | NP_057567.3 | |
XM_011533826.2 | 920 | Missense Mutation | AAT,AGT | N,S 54 | XP_011532128.1 |