Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGGACACCTTCCTTTCCCATGA[C/G]TATAACCAATGCTGGAGTACACAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176590 MIM: 609247 | ||||||||||||||||||||
Literature Links: |
LOC646903 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC646903 - uncharacterized LOC646903 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFN2 - profilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002628.4 | 757 | Missense Mutation | CTC,GTC | L,V 113 | NP_002619.1 | |
NM_053024.3 | 757 | UTR 3 | NP_444252.1 |
RNF13 - ring finger protein 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |